Genomic DNA isolation from tissues, NucleoSpin® 8/96 tissues/Tissue Core kits
NucleoSpin® 8/96 Tissue and NucleoSpin® 8/96 Tissue Core kits for isolation of genomic DNA from human or animal tissue, cultured cells, or bacteria. Available with a choice of flexible 8-well strip format for varying throughput or 96-well format for high throughput.
- Processing possible under vacuum or by centrifugation
- Suitable for manual and automated processing
- Innovative MN wash plate minimizes risk of cross-contamination
- DNA ready to use for, for example, PCR, Southern blotting, any kind of enzymatic reaction
Reduced kit composition together with a large variety of separately available accessories allow optimal adjustment of the kits to individual needs.
Specifications
Binding capacity: 40 µg
Elution volume: 100 - 200 µl
Sample size: <20 mg human/animal tissue, <106 human/animal cells, bacteria
Sample type: Bacterial cells;Human and animal fatty tissues
Time : 20 min/6 strips , 60 min/plate
: NucleoSpin® 8 Tissue includes NucleoSpin® Tissue Binding Strips, MN wash plate, MN square-well blocks, rack of tube strips, self-adhering foil, buffers, Proteinase K. NucleoSpin® 8 Tissue Core Kits include NucleoSpin® Tissue Binding Strips, buffers, Proteinase K.
NucleoSpin® 96 Tissue includes NucleoSpin® Tissue Binding Plates, MN wash plates, MN square-well blocks, round-well blocks, racks with tube strips, cap strips, self-adhering foil, buffers, Proteinase K. NucleoSpin® 8 Tissue Core Kits include NucleoSpin® Tissue Binding Plates, buffers, Proteinase K.
NucleoSpin® 96 Tissue includes NucleoSpin® Tissue Binding Plates, MN wash plates, MN square-well blocks, round-well blocks, racks with tube strips, cap strips, self-adhering foil, buffers, Proteinase K. NucleoSpin® 8 Tissue Core Kits include NucleoSpin® Tissue Binding Plates, buffers, Proteinase K.
Frequently Bought Together