Order Entry
Austria
ContactUsLinkComponent
CA WIP VOA standard (11 components)
CA WIP VOA standard (11 components)
Catalog # REST30236
Supplier:  Restek
CAS Number:  
undefined
CA WIP VOA standard (11 components)
Catalog # REST30236
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    2000 µg/ml each in P and T methanol
  • Pk:
    1 mL
  • Description:
    CA WIP VOA standard
  • Pk Info:
    1 ml/ampoulel

Specifications

About this item

For monitoring samples suspected of gasoline contamination requires laboratories to calibrate and report these compounds.

It consists of benzene, chlorobenzene, 1,2-dichlorobenzene, 1,3-dichlorobenzene, 1,4-dichlorobenzene, ethylbenzene, methyl tert-butyl ether, toluene, m-xylene, o-xylene, p-xylene.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.