Sulphur simulated distillation standard (SSDS)
Catalog # REST33049
Supplier: Restek
CAS Number:
Specifications
- Pk:1 mL
- Description:Sulfur simulated distillation standard (SSDS)
- Pk Info:1 ml amber ampoule
Specifications
About this item
The standard consists of ethanethiol, 1-propanethiol, 1-butanethiol, 1-pentanethiol, 1-hexanethiol, 1-heptanethiol, 3,5-dimethylbenzenethiol, 1-octanethiol, 1-nonanethiol, 1-decanethiol, 1-pentadecanethiolol, 1-hexadecanethiol and 1-octadecanethiol.