8270 calibration mix #2 (11 components)
Catalog # REST31619
Supplier: Restek
CAS Number:
Specifications
- Concentration:2000 μg/ml each in methylene chloride:methanol (85:15)
- Pk:1 mL
- Description:8270 calibration mix #2
- Pk Info:1 ml/ampoule
Specifications
About this item
Contains: Aniline (62-53-3); Benzidine (92-87-5); 4-Chloroaniline (106-47-8); 3,3'-Dichlorobenzidine (91-94-1); Diphenylamine (122-39-4)*; 2-Nitroaniline (88-74-4); 3-Nitroaniline (99-09-2); 4-Nitroaniline (100-01-6); N-Nitrosodimethylamine (62-75-9); N-Nitrosodi-n-propylamine (621-64-7); Pyridine (110-86-1)
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.