Order Entry
Austria
ContactUsLinkComponent
EPA method 8061A phthalate esters mixture (15 components)
EPA method 8061A phthalate esters mixture (15 components)
Catalog # REST33227
Supplier:  Restek
CAS Number:  
undefined
EPA method 8061A phthalate esters mixture (15 components)
Catalog # REST33227
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml each in hexane:acetone (80:20)
  • Pk:
    1 Ampoul
  • Description:
    EPA method 8061A phthalate esters mixture
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

The mixture consists of benzyl butyl phthalate, bisphthalate, bisphthalate, bisphthalate, bisphthalate, bisphthalate, di-n-butylphthalate, diethylphthalate, di-n-hexyl phthalate, dimethylphthalate, di-nonyl phthalate, di-n-octyl phthalate, dipentylphthalate, phthalic acid dicyclohexyl ester, phthalic acid diisobutyl ester.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.