Order Entry
Austria
ContactUsLinkComponent
CLP 04.1 BNA surrogate mix (8 components)
CLP 04.1 BNA surrogate mix (8 components)
Catalog # REST31493
Supplier:  Restek
CAS Number:  
undefined
CLP 04.1 BNA surrogate mix (8 components)
Catalog # REST31493
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    In methylene chloride
  • Pk:
    1 mL
  • Description:
    CLP 04.1 BNA Surrogate Mix
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

The mix consists of components such as 2-chlorophenol-d4 (1500 μg/ml), 1,2-dichlorobenzene-d4 (1000 μg/ml), 2-fluorobiphenyl (1000 μg/ml), 2-fluorophenol (1500 μg/ml), nitrobenzene-d5 (1000 μg/ml).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.