Order Entry
Austria
ContactUsLinkComponent
ASTM D3606 quality control standard with sec-butanol internal standard (3 components)
ASTM D3606 quality control standard with sec-butanol internal standard (3 components)
Catalog # REST30670
Supplier:  Restek
CAS Number:  
undefined
ASTM D3606 quality control standard with sec-butanol internal standard (3 components)
Catalog # REST30670
Supplier:  Restek
CAS Number:  

Specifications

  • Pk:
    25 mL
  • Description:
    ASTM D3606 quality control standard with sec-butanol internal standard, in isooctane
  • Pk Info:
    25 ml/ampoule

Specifications

About this item

Contains three components, those are benzene, 2-butanol (sec-butyl alcohol) and toluene.

Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.