Order Entry
Austria
ContactUsLinkComponent
DRO/ORO range calibration standard, (12 components)
DRO/ORO range calibration standard, (12 components)
Catalog # REST31832
Supplier:  Restek
CAS Number:  
undefined
DRO/ORO range calibration standard, (12 components)
Catalog # REST31832
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    2000 µg/ml each in methylene chloride
  • Pk:
    1 Ampoul
  • Description:
    DRO/ORO range calibration standard
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

This consists of n-decane (C10), n-dodecane (C12), n-tetradecane (C14), n-hexadecane (C16), n-octadecane (C18), n-eicosane (C20) ,n-docosane (C22) , n-tetracosane (C24), n-hexacosane (C26), n-octacosane (C28), n-triacontane (C30) and n-dotriacontane (C32).