ContactUsLinkComponent
NPI-1000 column test mix (8 components)
Catalog # REST35224
Supplier:  Restek
CAS Number:  
undefined
NPI-1000 column test mix (8 components)
Catalog # REST35224
Supplier:  Restek
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    NPI-1000 column test mix, 1000 µg/ml each in methylene chloride
  • Pk Info:
    1 ml/ampoule
Specifications
About this item
The mix consists of (C9:0) methyl nonanoate, (C13) n-tridecane, (C15) n-pentadecane, acenaphthylene, 4-chlorophenol, 1,6-hexanediol, 4-propylaniline and 1-undecanol.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.