Order Entry
Austria
ContactUsLinkComponent
Method 1664 oil and grease mix (2 components)
Artikelnr. REST31457
Lieferant:  Restek
CAS-Nummer:  
undefined
Method 1664 oil and grease mix (2 components)
Artikelnr. REST31457
Lieferant:  Restek
CAS-Nummer:  

Spezifikationen

  • Concentration:
    4000 μg/ml each in acetone
  • Pk:
    5 mL
  • Description:
    Method 1664 oil and grease mix
  • Pk Info:
    5 ml/ampoule

Spezifikationen

Über diesen Artikel

Contains two components, those are n-hexadecane and stearic acid.

Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.