Pesticide surrogate mix, EPA 8080, 8081, DCB and TCMX (2 components)
Catalog # REST32457
Supplier: Restek
CAS Number:
Specifications
- Concentration:200 μg/ml each in acetone
- Pk:1 mL
- Description:Pesticide surrogate mix, EPA 8080, 8081, DCB and TCMX
- Pk Info:1 ml/ampoule
Specifications
About this item
Contains two components, those are decachlorobiphenyl and 2,4,5,6-tetrachloro-m-xylene.
Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.