Organochlorine pesticide resolution check mix (with surrogates) (22 components)
Catalog # REST32454
Supplier: Restek
CAS Number:
Specifications
- Concentration:In hexane:toluene (90:10)
- Pk:1 mL
- Description:Organochlorine pesticide resolution check mix (with surrogates)
- Pk Info:1 ml/ampoule
Specifications
About this item
Contains 22 components, those are aldrin, α-bhc, β-bhc, δ-bhc, γ-bhc, cis-chlordane, trans-chlordane, decachlorobiphenyl, dieldrin, 4,4'-ddd, 4,4'-dde, 4,4'-ddt, endosulfan i, endosulfan ii, endosulfan sulfate, endrin, endrin aldehyde, endrin ketone, heptachlor, heptachlor epoxide, methoxychlor and 2,4,5,6-tetrachloro-m-xylene.
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.