ASTM D3606 calibration standard #4 without internal standard (2 components)
Catalog # REST30650
Supplier: Restek
CAS Number:
Specifications
- Pk:1 Ampoul
- Description:ASTM D3606 calibration standard #4 without internal standard, in isooctane
- Pk Info:25 ml/ampoule
Specifications
About this item
The mix consists of benzene (0,67% vol/vol), toluene (5% vol/vol).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.