Order Entry
Austria
ContactUsLinkComponent
ASTM D2887-12 calibration standard (20 components)
ASTM D2887-12 calibration standard (20 components)
Catalog # REST31674
Supplier:  Restek
CAS Number:  
undefined
ASTM D2887-12 calibration standard (20 components)
Catalog # REST31674
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    1% w/w in carbon disulphide
  • Pk:
    1
  • Description:
    ASTM D2887-12 calibration standard
  • Pk Info:
    1 g solution/ampoule

Specifications

About this item

Designed to serve as a column resolution text mixture, as well as a calibration mixture for retention time (RT) versus boiling point (bp) calibration.

The mix consists of, (C5) n-pentane, (C6) n-hexane, (C7) n-heptane, (C8) n-octane, (C9) n-nonane, (C10) n-decane, (C11) n-undecane, (C12) n-dodecane, (C14) n-tetradecane, (C15) n-pentadecane, (C16) n-hexadecane, (C17) n-heptadecane, (C18) n-octadecane, (C20) n-eicosane, (C24) n-tetracosane, (C28) n-octacosane, (C32) n-dotriacontane, (C36) n-hexatriacontane, (C40) n-tetracontane, (C44) n-tetratetracontane.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.