Order Entry
Austria
ContactUsLinkComponent
Organochlorine pesticide system evaluation mix (2 components)
Organochlorine pesticide system evaluation mix (2 components)
Catalog # REST32417
Supplier:  Restek
CAS Number:  
undefined
Organochlorine pesticide system evaluation mix (2 components)
Catalog # REST32417
Supplier:  Restek
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Description:
    Organochlorine pesticide system evaluation mix, in methyl tert-butyl ether
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Contains two components, those are 4,4'-DDT and endrin.

  • Designed for daily assessment of system performance
  • Reveals active sites in the injection port and/or GC column

Prepared in MTBE—low expansion volume helps minimise backflash.

Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.