Orders LinkContactUsLinkComponent
XERT-500 column test mix (8 components)
XERT-500 column test mix (8 components)
XERT-500 column test mix, 500 µg/ml each in toluene, 1 ml/ampoule, 1
Catalog # REST35247
Supplier:   Restek
XERT-500 column test mix (8 components)
Catalog # REST35247
Supplier:  Restek
CAS Number:  
XERT-500 column test mix (8 components)
XERT-500 column test mix (8 components)
XERT-500 column test mix, 500 µg/ml each in toluene, 1 ml/ampoule, 1
CAS Number:  
Price
64,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
This contains (C9:0 FAME) methyl nonanoate, (C13) n-tridecane, (C15) n-pentadecane, acenaphthylene, 4-chlorophenol, n-decylamine, 1,6-hexanediol and 1-undecanol (112-42-5).
Specifications
Specifications
  • Pk:
    1
  • Description:
    XERT-500 column test mix, 500 µg/ml each in toluene
  • Pk Info:
    1 ml/ampoule