Order Entry
Austria
ContactUsLinkComponent
Extraction retention time standard, (4 components)
Extraction retention time standard, (4 components)
Catalog # REST31830
Supplier:  Restek
CAS Number:  
undefined
Extraction retention time standard, (4 components)
Catalog # REST31830
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    2000 µg/ml each in methylene chloride
  • Pk:
    1 Ampoul
  • Description:
    Extraction retention time standard
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

This contains n-hexane (C6), n-decane (C10), n-docosane (C22) and n-dotriacontane (C32).