ContactUsLinkComponent
EPA 539 calibration stock standard (7 components)
EPA 539 calibration stock standard (7 components)
Catalog # REST31998
Supplier:  Restek
CAS Number:  
undefined
EPA 539 calibration stock standard (7 components)
Catalog # REST31998
Supplier:  Restek
CAS Number:  
Specifications
  • Pk:
    1 Ampoul
  • Description:
    EPA 539 calibration stock standard
  • Pk Info:
    1 ml/ampoule
Specifications
About this item
Contains the seven hormones listed in EPA Method 539 at the appropriate concentrations. The seven components are 4-androstene-3, 17-dione, equilin, 17-beta-Estradiol, estriol, estrone, 17-alpha-ethynylestradiol and testosterone.
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Ordering information: UCMR3 method 539 calibration standard with concentrations of 10 to 400 µg/ml to more conveniently match UCMR3 requirements (cat.# 32461) also available.