Order Entry
Austria
ContactUsLinkComponent
SV calibration mix #5/610 PAH mix (16 components)
SV calibration mix #5/610 PAH mix (16 components)
Catalog # REST31011
Supplier:  Restek
CAS Number:  
undefined
SV calibration mix #5/610 PAH mix (16 components)
Catalog # REST31011
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    2000 μg/ml each in methylene chloride
  • Pk:
    1
  • Description:
    SV calibration mix #5/610 PAH mix
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

SV calibration mix contains acenaphthene, acenaphthylene, anthracene, benz(a)anthracene, benzo(a)pyrene, benzo(b)fluoranthene, benzo(k)fluoranthene, benzo(ghi)perylene, chrysene, dibenz(a,h)anthracene, fluoranthene, fluorene, indeno(1,2,3-cd)pyrene, naphthalene, phenanthrene, pyrene.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.