Order Entry
Austria
ContactUsLinkComponent
SV system performance check mix (4 components)
SV system performance check mix (4 components)
Catalog # REST31689
Supplier:  Restek
CAS Number:  
undefined
SV system performance check mix (4 components)
Catalog # REST31689
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    2000 µg/ml each in methylene chloride
  • Pk:
    1 Ampoul
  • Description:
    SV system performance check mix (SPCC)
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

This SV system performance check mix consist of four components: 2,4-Dinitrophenol, Hexachlorocyclopentadiene, 4-Nitrophenol and N-Nitroso-di-n-propylamine.

Shelf life 6 to 24 months.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.