- Concentration:1000 µg/ml each in methylene chloride (3-methylphenol and 4-methylphenol at 500 µg/ml)
- Pk:1 Ampoul
- Description:SOM01.1 SVOA MegaMix standard, revised
- Pk Info:1 ml/ampoule
Consists of 65 components, those are acenaphthene, acenaphthylene, acetophenone, anthracene, benzanthracene, benzopyrene, benzofluoranthene, benzoperylene, benzofluoranthene, benzyl butyl phthalate, biphenyl, bismethane, bisether, bisphthalate, 4-bromophenyl-phenylether, carbazole, 4-chloroaniline, 4-chloro-3-methylphenol, 2-chloronaphthalene, 2-chlorophenol, 4-chlorophenyl-phenylether, chrysene, dibenzanthracene, dibenzofuran, 3,3'-dichlorobenzidine, 2,4-dichlorophenol, diethylphthalate, 2,4-dimethylphenol, dimethylphthalate, di-n-butylphthalate, 4,6-dinitro-2-methylphenol , 2,4-dinitrophenol, 2,4-dinitrotoluene, 2,6-dinitrotoluene, di-n-octyl phthalate, diphenylamine *,fluoranthene, fluorene, hexachlorobenzene, hexachloro-1,3-butadiene , hexachlorocyclopentadiene, hexachloroethane, indenopyrene, isophorone, 2-methylnaphthalene, 2-methylphenol , 3-methylphenol, 4-methylphenol , naphthalene, 2-nitroaniline, 3-nitroaniline, 4-nitroaniline, nitrobenzene, 2-nitrophenol, 4-nitrophenol, n-nitroso-di-n-propylamine, 2,2'-oxybis, pentachlorophenol, phenanthrene, phenol, pyrene, 1,2,4,5-tetrachlorobenzene, 2,3,4,6-tetrachlorophenol, 2,4,5-trichlorophenol and 2,4,6-trichlorophenol.