System performance test standard mixture of n-Alkanes (16 components)
Catalog # REST31678
Supplier: Restek
CAS Number:
Specifications
- Concentration:50 µg/ml each in hexane
- Pk:1
- Description:System performance test standard mixture of n-Alkanes
- Pk Info:1 ml/ampoule
Specifications
About this item
This standard mixture cobtains: (C10) n-Decane, (C12) n-Dodecane, (C14) n-Tetradecane, (C16) n-Hexadecane, (C18) n-Octadecane, (C20) n-Eicosane, (C22) n-Docosane, (C24) n-Tetracosane, (C26) n-Hexacosane, (C28) n-Octacosane, (C30) n-Triacontane, (C32) n-Dotriacontane, (C34) n-Tetratriacontane, (C36) n-Hexatriacontane, (C38) n-Octatriacontane and (C40) n-Tetracontane.
Shelf life 6 to 96 months.