Order Entry
Austria
ContactUsLinkComponent
Test mix E1618 (13 components)
Test mix E1618 (13 components)
Catalog # REST31613
Supplier:  Restek
CAS Number:  
undefined
Test mix E1618 (13 components)
Catalog # REST31613
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    0,05% volume/volume each in methylene chloride
  • Pk:
    1 mL
  • Description:
    E1618 test mix
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Components in this mix (0,5 μl/ml or 0,05% volume/volume each) are at 10X the concentration of the final test solution specified in ASTM 1618 and ASTM 1387.

The mix consists of (C6) n-hexane, (C8) n-octane, (C10) n-decane, (C12) n-dodecane, (C14) n-tetradecane, (C16) n-hexadecane, (C18) n-octadecane, (C20) n-eicosane, 2-ethyltoluene, 3-ethyltoluene, toluene, 1,2,4-trimethylbenzene, p-xylene.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.