E1387 column resolution check mix (13 components)
Catalog # REST31224
Supplier: Restek
CAS Number:
Specifications
- Concentration:2000 µg/ml each in methylene chloride
- Pk:1
- Description:E1387 olumn resolution check mix
- Pk Info:1 ml/ampoule
Specifications
About this item
This mix also can be used for underground storage tank monitoring.
It consist of 13 components: (C6) n-Hexane, (C8) n-Octane, (C10) n-Decane, (C12) n-Dodecane, (C14) n-Tetradecane, (C16) n-Hexadecane, (C18) n-Octadecane, (C20) n-Eicosane, 2-Ethyltoluene, 3-Ethyltoluene, Toluene, 1,2,4-Trimethylbenzene and p-Xylene.
Shelf life maximum 85 months and minimum 6 months.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.