Steroids and mixed pharmaceuticals mix (10 components)
REST31117
: Restek
:
- Concentration:200 µg/ml each in acetonitrile
- Pk:1 Ampoul
- Description:Steroids and mixed pharmaceuticals mix
- Pk Info:1 ml/ampoule
This steroids and mixed pharmaceuticals mix contain ten components: Bisphenol A, Diclofenac sodium salt, 17-beta-Estradiol, Estrone, 17-alpha-Ethynylestradiol, 4-para-Nonylphenol, 4-tert-Octylphenol, Primidone, Progestrone and Testosterone.
Shelf life 24 months.
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.