Order Entry
Austria
ContactUsLinkComponent
508.1 calibration mix #3 (8 components)
508.1 calibration mix #3 (8 components)
Catalog # REST32096
Supplier:  Restek
CAS Number:  
undefined
508.1 calibration mix #3 (8 components)
Catalog # REST32096
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    500 µg/ml each in ethyl acetate
  • Pk:
    1
  • Description:
    508.1 calibration mix #3
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Consists of eight components, those are alachlor, atrazine, chlorothalonil, cyanazine , hexachlorocyclopentadiene, metolachlor, metribuzin and simazine.

8 components

Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.