Order Entry
Austria
Orders LinkContactUsLinkComponent
MOSH/MOAH standard (9 components) | Avantor

MOSH/MOAH standard (9 components)

MOSH/MOAH standard, 150 - 600 µg/ml each in toluene, 1 ml/ampoule, 1
Catalog # REST89031070
Supplier:   Restek
MOSH/MOAH standard (9 components)
Catalog # REST89031070
Supplier:  Restek
CAS Number:  
MOSH/MOAH standard (9 components)
MOSH/MOAH standard (9 components)

MOSH/MOAH standard, 150 - 600 µg/ml each in toluene, 1 ml/ampoule, 1

CAS Number:  
Price
66,69 €
Log in for availability
1 * 1 Pack.
Product Details & Documents

This MOSH/MOAH standard developed specifically for using LC coupled with GC-FID to accurately determine hydrocarbons found in mineral oils, which can migrate from packaging into food.

This mix contains non-interfering internal standards as well as both mineral oil saturated hydrocarbon (MOSH) and mineral oil aromatic hydrocarbon (MOAH) markers to correctly cut fractions.

It contains nine components: (C11) n-Undecane (300 μg/ml), (C13) n-Tridecane (150 μg/ml), Bicyclohexyl (300 μg/ml), Cholestane (5-alpha-cholestane) (600 μg/ml), 1-Methylnaphthalene (300 μg/ml), 2-Methylnaphthalene (300 μg/ml), n-Pentylbenzene (300 μg/ml), Perylene (600 μg/ml) and 1,3,5-Tri-tert-butylbenzene (300 μg/ml).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.

Specifications

Specifications

  • Concentration:
    150 - 600 µg/ml each in toluene
  • Pk:
    1
  • Description:
    MOSH/MOAH standard
  • Pk Info:
    1 ml/ampoule

Specifications