Pesticide performance evaluation mix with surrogates (8 components)
Catalog # REST32074
Supplier: Restek
CAS Number:
Specifications
- Concentration:In hexane
- Pk:1 mL
- Description:Pesticide performance evaluation mix
- Pk Info:1 ml/ampoule
Specifications
About this item
This mix contains eight components.
Components are α-BHC (1 μg/ml), β-BHC (1 μg/ml), γ-BHC (1 μg/ml), 4,4'-DDT (10 μg/ml), decachlorobiphenyl (SS) (2 μg/ml), endrin (5 μg/ml), methoxychlor (25 μg/ml), 2,4,5,6-tetrachloro-m-xylene (2 μg/ml).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.