NJDEP EPH 10/08 rev. 2 aromatics fractionation check mix (18 components)
Catalog # REST30545
Supplier: Restek
CAS Number:
Specifications
- Concentration:400 µg/ml each in hexane:toluene (50:50)
- Pk:5 mL
- Description:NJDEP EPH 10/08 rev.2 aromatics fractionation Check mix
- Pk Info:5 ml/ampoule
Specifications
About this item
Consists of 18 components, those are acenaphthene, acenaphthylene, anthracene, benzanthracene, benzopyrene, benzofluoranthene, benzoperylene, benzofluoranthene, chrysene, dibenzanthracene, fluoranthene, fluorene, indenopyrene, 2-methylnaphthalene, naphthalene, phenanthrene, 1,2,3-trimethylbenzene and pyrene
16 components
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.