Chloroprene
REST30238
: Restek
:
- Concentration:5,000 µg/ml in P and T methanol
- Pk:1 mL
- Description:Chloroprene
- Pk Info:1 ml/ampoule
A pure, quantitative chloroprene solution, specially stabilised in purge-and-trap grade methanol to allow dilutions for working standards in unmodified purge-and-trap methanol. the entire preparation procedure is performed under carefully monitored conditions to prevent any contamination or polymerisation of the highly reactive, neat chloroprene.
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.