Order Entry
Austria
ContactUsLinkComponent
MA VPH matrix spike mix with surrogate (14 components)
MA VPH matrix spike mix with surrogate (14 components)
Catalog # REST30454
Supplier:  Restek
CAS Number:  
undefined
MA VPH matrix spike mix with surrogate (14 components)
Catalog # REST30454
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    2500 µg/ml each in P and T methanol
  • Pk:
    1 mL
  • Description:
    MA VPH matrix spike mix with surrogate
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

The MA VPH matrix spike mix consists of 14 components they are (C5) n-pentane, (C9) n-nonane, benzene, 2,5-dibromotoluene (SS), ethylbenzene, 2-methylpentane, methyl tert-butyl ether (mtbe), naphthalene, toluene, 1,2,4-trimethylbenzene, 2,2,4-trimethylpentane (isooctane), m-xylene, o-xylene and p-xylene.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.