Order Entry
Austria
ContactUsLinkComponent
California oxygenates mix (5 components)
California oxygenates mix (5 components)
Catalog # REST30465
Supplier:  Restek
CAS Number:  
undefined
California oxygenates mix (5 components)
Catalog # REST30465
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    In P and T methanol
  • Pk:
    1 mL
  • Description:
    California oxygenates mix
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

The california oxygenates mix consists of five components they are tert-amyl methyl ether (TAME) 2000 μg/ml, tert-butanol (TBA) 10000 μg/ml, diisopropyl ether (DIPE) 2000 μg/ml, ethyl-tert-butyl ether (ETBE) 2000 μg/ml and methyl tert-butyl ether (MTBE) 2000 μg/ml.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.