606 Phthalate esters calibration mix (6 components)
Catalog # REST31031
Supplier: Restek
CAS Number:
Specifications
- Concentration:2000 µg/ml each in methanol
- Pk:1 mL
- Description:606 Phthalate Esters Calibration Mix
- Pk Info:1 ml/ampoule
Specifications
About this item
Calibration mix contains Bis(2-ethylhexyl)phthalate, benzyl butyl phthalate, di-n-butyl phthalate, diethylphthalate, dimethylphthalate, di-n-octyl phthalate.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.