Pesticide reference standard, Trifluralin
REST32238
: Restek
:
- Concentration:1000 μg/ml in methanol
- Pk:1
- CAS No.:1582-09-8
- Description:Trifluralin
- Formula:C₁₃H₁₆F₃N₃O₄
- Description:Trifluralin
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.