Method 525.2 internal standard mix
Catalog # REST31825
Supplier: Restek
CAS Number:
Specifications
- Concentration:1000 µg/ml each in acetone
- Pk:1 Ampoul
- Description:Method 525.2 internal standard mix
- Pk Info:1 ml
Specifications
About this item
This mix contains 3 components: acenaphthene-d10, chrysene-d12 and phenanthrene-d10.
Certifications: Certified reference materials (CRMs) manufactured and QC tested in ISO-accredited labs satisfy your ISO requirements.