8270/Appendix IX kit
Catalog # REST31815
Supplier: Restek
CAS Number:
Specifications
- Pk:5
- Description:8270/Appendix IX kit
- Pk Info:5 mixtures (5×1 ml)
Specifications
About this item
This kit contains 1 ml each of these mixtures: 8270 MegaMix standard, 605 benzidines calibration mix, appendix IX mix #1, revised (methapyrilene is not included in this revised standard), methapyrilene standard, appendix IX Mix #2.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.