Express Eingabe
Austria
ContactUsLinkComponent
Method 525.2 semivolatile mix (revised)
Method 525.2 semivolatile mix (revised)
Catalog # REST31899
Supplier:  Restek
CAS Number:  
undefined
Method 525.2 semivolatile mix (revised)
Catalog # REST31899
Supplier:  Restek
CAS Number:  
Specifications
  • VE:
    1 Ampoul
  • Bezeichnung:
    Method 525.2 semivolatile mix (revised), 1000 µg/ml each in acetone (pentachlorophenol at 4000 µg/ml)
  • Pk Info:
    1 ml/ampoule
Specifications
About this item
This method 525.2 semivolatile mix contain 28 components.