Order Entry
Austria
ContactUsLinkComponent
EPA method 8310 PAH mixture
EPA method 8310 PAH mixture
Catalog # REST31874
Supplier:  Restek
CAS Number:  
undefined
EPA method 8310 PAH mixture
Catalog # REST31874
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    500 µg/ml each
  • Pk:
    1 mL
  • Description:
    EPA method 8310 PAH mixture
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

EPA method 8310 PAH mixturecontain 18 components.

Matrix: Acetonitrile:toluene (92:8)

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.