LC OQ standards kit
Catalog # REST31069
Supplier: Restek
CAS Number:
Specifications
- Pk:1
- Description:LC OQ standards kit
Specifications
About this item
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Delivery: Kit contains LC OQ gradient standard (1 ml), LC OQ wavelength accuracy standard (5 ml) and LC OQ linearity kit (6×1 ml ampoule).