FID performance evaluation standard
Catalog # REST33908
Supplier: Restek
CAS Number:
Specifications
- Pk:1 mL
- Description:FID performance evaluation standard, 0,03% w/w each in hexane
- Pk Info:1 ml/ampoule
Specifications
About this item
FID performance evaluation standard contains three components such as (C14) n-Tetradecane (629-59-4), (C15) n-Pentadecane (629-62-9) and (C16) n-Hexadecane (544-76-3).
Certifications: Certified reference materials (CRMs) manufactured and QC tested in ISO accredited labs satisfy ISO requirements.