Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Chromatography Sorbents
HPLC Sorbents
Bulk packing, PWXL Top-Off
Bulk packing, PWXL Top-Off
Bulk packing, PWXL Top-Off
Catalog # TOYO0008035
Supplier:
TOSOH Bioscience
CAS Number:
Expand image
Bulk packing, PWXL Top-Off
Catalog # TOYO0008035
Supplier:
TOSOH Bioscience
CAS Number:
Specifications
Particle Size:
10 µm
Pk:
1 g
Description:
Bulk packing, PWXL Top-Off, 1 g wet resin
Specifications
About this item
Product Details
Product Details