Revised GPC calibration mix, semi volatile compounds
Supplier: Restek
Certificates
About this item
Revised GPC calibration mix is a qualitative mixture and it is useful for determining GPC dump/collect times. The compounds are dissolved in methylene chloride at various concentrations.
It is made up of five components: 5 mg/ml of Bis(2-ethylhexyl)phthalate, 250 mg/ml of corn oil, 1,0 mg/ml of methoxychlor, 0,2 mg/ml of perylene, 0,8 mg/ml of sulphur.
Frequently Bought Together