ContactUsLinkComponent
AiroSensor X ER 20-20-43 Wireless Ultra-low Temperature Data Logger (−200 to +200 °C)
AiroSensor X ER 20-20-43 Wireless Ultra-low Temperature Data Logger (−200 to +200 °C)
Supplier:  SenseAnywhere
Certificates
undefined
AiroSensor X ER 20-20-43 Wireless Ultra-low Temperature Data Logger (−200 to +200 °C)
Supplier:  SenseAnywhere
Specifications
  • Catalog No:
  • 620-2701
  • SENA0096647494807
  • Model:
  • AiroSensor X ER model 20-20-43
  • AiroSensor X ER model 20-20-43
  • Protection class:
  • IP 67
  • IP 67
  • Battery life:
  • 10 years
  • 10 years
  • Weight:
  • 56 g
  • 56 g
  • W×D×H:
  • 35×25×143 mm
  • 35×25×143 mm
  • Data memory - No. of readings:
  • 1000000
  • 1000000
Specifications
Related Information
SAB temperature PT100 probe
Product Information
Frequently Bought Together