Order Entry
Austria
ContactUsLinkComponent
Certified aromatics in gasoline, 16 components
Certified aromatics in gasoline, 16 components
Catalog # REST30485
Supplier:  Restek
CAS Number:  
Certified aromatics in gasoline, 16 components
Catalog # REST30485
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    5500 ppm gasoline in P and T methanol
  • Pk:
    1
  • Description:
    Certified aromatics in gasoline
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Confirms unleaded gasoline TPH, BTEX and aromatics in single analysis.

This standard consists of 16 components.

Components: Benzene, ethylbenzene, m-ethyltoluene, o-ethyltoluene, p-ethyltoluene, isopropylbenzene, methyl tert-butyl ether (MTBE), naphthalene, n-propylbenzene, toluene, 1,2,3-trimethylbenzene, 1,2,4-trimethylbenzene, 1,3,5-trimethylbenzene, m-xylene, o-xylene, p-xylene.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.