Order Entry
Austria
ContactUsLinkComponent
Gasoline component standard, 10 components
Gasoline component standard, 10 components
Catalog # REST30486
Supplier:  Restek
CAS Number:  
Gasoline component standard, 10 components
Catalog # REST30486
Supplier:  Restek
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Description:
    Gasoline component standard, 10000 µg/ml total in P and T methanol
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.

Gasoline component standard contains 10 components.

Components: Benzene (500 µg/ml), ethylbenzene (500 µg/ml), heptane (500 µg/ml), 2-methylpentane (1500 µg/ml), toluene (1500 µg/ml), 1,2,4-trimethylbenzene (1500 µg/ml), 2,2,4-trimethylpentane (1500 µg/ml), m-xylene (1000 µg/ml), o-xylene (1000 µg/ml) and p-xylene (1000 µg/ml).