Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Food Standards
FAME mix in rapeseed oil
FAME mix in rapeseed oil
FAME mix in rapeseed oil
Catalog # HEWL5191-4277
Supplier:
Agilent
CAS Number:
Expand image
FAME mix in rapeseed oil
Catalog # HEWL5191-4277
Supplier:
Agilent
CAS Number:
Specifications
Pk:
1
Description:
FAME mix in rapeseed oil, 1 ml
Specifications
About this item
Product Details
Product Details