LC OQ standards kit
Catalog # REST30012
Supplier: Restek
CAS Number:
Specifications
- Pk:1 mL
- Description:LC OQ standards kit (acetone)
Specifications
About this item
Contains: LC OQ gradient standard (1 ml), LC OQ wavelength accuracy standard (5 ml), LC OQ linearity kit (six 1 ml/ampoule).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Ordering information: Components of the LC linearity kit are not available for individual sale.