
Hydrochloric acid concentrate for dilution to 1 L, 0.1 M HCl (0.1 N) volumetric solution, Fluka™
:7647-01-0
:
:HCl
:MFCD00011324
:100 °C (1.013 hPa)
:1.030 g/cm³ (20 °C)
:1789
:8,III
:36.46 g/mol
:GHS05
:Ambient
- Factor:0.998 - 1.002
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective
Frequently Bought Together