Ferroin (1,10-phenanthrolineferrous sulphate complex) 0.025 M in aqueous solution redox indicator, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:14634-91-4
:
Formula:C₃₆H₂₄FeN₆SO₄
MDL Number:MFCD00036428
:692.54 g/mol
Boiling Pt:100 °C (solvent)
Melting Pt:0 °C
Density:1.006 g/cm³
Flash Pt: Does not flash
Storage Temperature:Ambient
Specification Test Results
- Appearance of the substance:complying
- Density (D 20/4):1.003 - 1.008
- Suitability as indicator:complying
Specification Test Results
Frequently Bought Together