Order Entry
Austria
ContactUsLinkComponent
Magnesium hydrogen orthophosphate trihydrate ≥98%, Puriss. special, meets analytical specification of DAB, Fluka™
Magnesium hydrogen orthophosphate trihydrate ≥98%, Puriss. special, meets analytical specification of DAB, Fluka™
Catalog # 04260-30KG
CAS Number:  
Magnesium hydrogen orthophosphate trihydrate ≥98%, Puriss. special, meets analytical specification of DAB, Fluka™
Catalog # 04260-30KG
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    30 kg
  • Seq:
    4319
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay:
    98 - 103 %
  • Barium (Ba):
    complying
  • Arsenic (As):
    Max. 0.0001 %
  • Iron (Fe):
    Max. 0.005 %
  • Heavy metals (as Pb):
    Max. 0.002 %
  • Chloride (Cl):
    Max. 0.02 %
  • Sulfate (SO₄):
    Max. 0.03 %
  • Mg-, Mg-dihydrogen phosphate:
    complying

Specification Test Results

About this item

Synonyms: Magnesium hydrogen phosphate trihydrate, Magnesium phosphate dibasic trihydrate
 :
 :174.33 g/mol
Melting Pt:550 °C
Density:2.1 g/cm3 (20 °C)
Storage Temperature:Ambient
EINECS:233-142-9
CAS Number:7782-75-4
Formula:HMgO₄P·3H₂O
MDL Number:MFCD00150390